Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
3-Methoxy-2,6-dimethylaniline 1 * 50 g
Catalog # MSPPAG020L84-50G
Supplier:
MARKET SOURCE PART PROCESS
Expand image
3-Methoxy-2,6-dimethylaniline 1 * 50 g
Catalog # MSPPAG020L84-50G
Supplier:
MARKET SOURCE PART PROCESS
Frequently Bought Together
N-Ethyldiisopropylamine ≥99.5% for peptide synthesis
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Petroleum spirit, 60…95 °C, TECHNICAL
Supplier: VWR Chemicals