Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
(SP-4-1)-[1,3-BIs[2,6-bis(1-ethylpropyl)phenyl]-4,5-dichloro-1,3-dihydro-2H-imidazol-2-ylidene]dichloro(2-methylpyridine)palladium 1 * 5 g
Catalog # MSPPAG01X9FB-5G
Supplier:
MARKET SOURCE PART PROCESS
Expand image
(SP-4-1)-[1,3-BIs[2,6-bis(1-ethylpropyl)phenyl]-4,5-dichloro-1,3-dihydro-2H-imidazol-2-ylidene]dichloro(2-methylpyridine)palladium 1 * 5 g
Catalog # MSPPAG01X9FB-5G
Supplier:
MARKET SOURCE PART PROCESS
Frequently Bought Together
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water with 0,1% (v/v) formic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Dimethyl sulphoxide ≥99.9%, ChromAR® for HPLC, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Pyridine, anhydrous (max. 0.003% H₂O) ≥99.8%
Supplier: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Benzyl alcohol 98.0-100.5% NF, Multi-Compendial low in peroxide, J.T.Baker®
Supplier: Avantor