Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
D-Lin-MC3-DMA 1 * 5 g
MSPPAG01EO59-5G
:
MARKET SOURCE PART PROCESS
D-Lin-MC3-DMA 1 * 5 g
MSPPAG01EO59-5G
:
MARKET SOURCE PART PROCESS
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Petroleum spirit, 40…60 °C, AnalaR NORMAPUR® ACS analytical reagent (max. 0.01% aromatic hydrocarbons)
: VWR Chemicals
Methanol, anhydrous (max. 0.002% H₂O) ≥99.9%
: VWR Chemicals
, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals