Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Methanesulfonato(diadamantyl-n-butylphosphino)-2'-amino-1,1'-biphenyl-2-yl)palladium(II) dichloromethane adduct 1 * 100 g
Catalog # MSPPAG01EGAC-100G
Supplier:
MARKET SOURCE PART PROCESS
Expand image
Methanesulfonato(diadamantyl-n-butylphosphino)-2'-amino-1,1'-biphenyl-2-yl)palladium(II) dichloromethane adduct 1 * 100 g
Catalog # MSPPAG01EGAC-100G
Supplier:
MARKET SOURCE PART PROCESS
Frequently Bought Together
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM®, super gradient grade for HPLC, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals