tert-butyl 2-bromo-6,7-dihydro-4H-[1,3]thiazolo[5,4-c]pyridine-5-carboxylate 1 * 100 g
Catalog # MSPPAG00I791-100G
Supplier: MARKET SOURCE PART PROCESS
tert-butyl 2-bromo-6,7-dihydro-4H-[1,3]thiazolo[5,4-c]pyridine-5-carboxylate 1 * 100 g
Catalog # MSPPAG00I791-100G
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Cyclohexane ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methyl ethyl ketone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Dichloromethane ≥99.8% stabilised, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals