Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Pyridine, 3-bromo-2-fluoro-6-methyl- 1 * 10 g
Catalog # MSPPAG00I7E9-10G
Supplier:
MARKET SOURCE PART PROCESS
Expand image
Pyridine, 3-bromo-2-fluoro-6-methyl- 1 * 10 g
Catalog # MSPPAG00I7E9-10G
Supplier:
MARKET SOURCE PART PROCESS
Frequently Bought Together
N,N-Dimethylformamide, Reagent Grade
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals