Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
L-Serine, N-[(1,1-dimethylethoxy)carbonyl]-, 1,1-dimethylethyl ester 1 * 100 g
Catalog # MSPPAG00G373-100G
Supplier:
MARKET SOURCE PART PROCESS
Expand image
L-Serine, N-[(1,1-dimethylethoxy)carbonyl]-, 1,1-dimethylethyl ester 1 * 100 g
Catalog # MSPPAG00G373-100G
Supplier:
MARKET SOURCE PART PROCESS
Frequently Bought Together
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals