0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Pump Tubing, GORE STA-PURE Series PCS, 0.8 mm ID x 4.0 mm OD 305 mm L 1 * 1 items
Catalog # MFLX96251-18
Supplier:
Avantor Fluid Handling
New Product
Expand image
Pump Tubing, GORE STA-PURE Series PCS, 0.8 mm ID x 4.0 mm OD 305 mm L 1 * 1 items
Catalog # MFLX96251-18
Supplier:
Avantor Fluid Handling
New Product
Frequently Bought Together
High-Density Polyethylene (HDPE) Tubing
Supplier: Avantor Fluid Handling
VWR® Desiccator Plates
Supplier: VWR Collection
Chemistry scoops
Supplier: BURKLE
VWR®, Funnels, Powder
Supplier: VWR Collection
TBNG NORPRENE BAR 1/8''ID 50' 1 * 50 Feet
Supplier: Avantor Fluid Handling
VWR®, Pestles
Supplier: VWR Collection
Tubing clamps, PVDF, LaboPlast®
Supplier: BURKLE
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, PharMed® BPT, Avantor®
Supplier: Avantor Fluid Handling