Order Entry
India
Orders LinkContactUsLinkComponent
VWR® Filter Papers, Qualitative and Quantitative

VWR® Filter Papers, Qualitative and Quantitative

S1, 90 mm Ø
Catalog # 516-1187
Supplier:   VWR Collection
VWR® Filter Papers, Qualitative and Quantitative
Catalog # 516-1187
Supplier:  VWR Collection
VWR® Filter Papers, Qualitative and Quantitative

S1, 90 mm Ø

Price
Inquire For Price
Log in for availability
1 * 100 items
Product Details & Documents

VWR® Filter papers offer a wide range of ash-less filter paper for qualitative and quantitative analysis.

  • Made of 100 % cotton linters
  • Ash-free (ash content ≤0,01% according to DIN 54370)
  • Wet-strengthened apart from S1 grade
  • Colour-coded box for easy selection
  • Supplied in rolls, sheets, discs, and folded filters

These qualitative filter papers are essentially used for analytical purposes and routine analyses, whenever no gravimetric analyses are required. They are wet-strengthened and can be used for pressure and vacuum filtration. They are made of refined pulp and linters with an >95% α-cellulose content, are very pure with an ash content <0,1%.

These filter papers are used for quantitative and gravimetric analyses as well as for pressure or vacuum filtration. They are made out of 100% cotton linters with an α-cellulose content of >98% and are acid-washed to make the papers ashless and achieve high purity.

Specifications
  • Grade:
    S1
  • Seq:
    01
  • Size:
    90 mm Ø