Order Entry
India
ContactUsLinkComponent
VWR STERILE BLUE SMS SLEEVE PROTECTOR, STANDARD SIZE, GAMMA STERILIZED, TURNED ENTRY CUFF, INDIVIDUALLY BAGGED IN PEEL / TEAR BAG, TUNNELIZED ELASTIC CUFFS, ULTRASONICALLY WELDED CONSTRUCTION, 200 EACH PER CASE, DOUBLE BAGGED CASE LINED, MADE IN U.S. 1 * 200 items
  VWRI76944-184
VWR STERILE BLUE SMS SLEEVE PROTECTOR, STANDARD SIZE, GAMMA STERILIZED, TURNED ENTRY CUFF, INDIVIDUALLY BAGGED IN PEEL / TEAR BAG, TUNNELIZED ELASTIC CUFFS, ULTRASONICALLY WELDED CONSTRUCTION, 200 EACH PER CASE, DOUBLE BAGGED CASE LINED, MADE IN U.S. 1 * 200 items
  VWRI76944-184
Frequently Bought Together