Order Entry
India
ContactUsLinkComponent
384-well Black polystyrene, glass bottomed with lid Individually packed. Sterile, case of 10 175µm borosilicate flat glass plate with high light transmission down to 335nm Tissue culture treated and Irradiated 1 * 10 items
Catalog # 732-3990
Supplier:  VWR Collection
384-well Black polystyrene, glass bottomed with lid Individually packed. Sterile, case of 10 175µm borosilicate flat glass plate with high light transmission down to 335nm Tissue culture treated and Irradiated 1 * 10 items
Catalog # 732-3990
Supplier:  VWR Collection
Frequently Bought Together