Order Entry
India
ContactUsLinkComponent
96-well Black polystyrene, glass bottomed with lid Individually packed. Sterile, case of 10 175µm borosilicate flat glass plate with high light transmission down to 335nm. Tissue culture treated and Irradiated 1 * 10 items
Catalog # 732-3989
Supplier:  VWR Collection
96-well Black polystyrene, glass bottomed with lid Individually packed. Sterile, case of 10 175µm borosilicate flat glass plate with high light transmission down to 335nm. Tissue culture treated and Irradiated 1 * 10 items
Catalog # 732-3989
Supplier:  VWR Collection