Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
HDPE BOTL RECT 2500 ML, CAP RED ST 1 * 35 items
Catalog # 215-4651
Supplier:
VWR Collection
Certificates
Expand image
HDPE BOTL RECT 2500 ML, CAP RED ST 1 * 35 items
Catalog # 215-4651
Supplier:
VWR Collection
Certificates
Related Information
Documentation
Datasheet
TDS HDPE Bottle.pdf
VWRI215-4651_2311070883.pdf
VWRI215-4651_2411251127.pdf
Documentation
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Platform Scales
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Sodium hydrogen carbonate 99.0-101.0% Ph. Eur.
Supplier: VWR Chemicals
VWR®, Bottles, HDPE
Supplier: VWR Collection