Order Entry
India
Orders LinkContactUsLinkComponent
 

VWR® PHP Plates

PHP Petri plate, stack size 10, 55 mm, Gamma irradiated, 10×112
  391-0684
VWR® PHP Plates
  391-0684
VWR® PHP Plates
VWR® PHP Plates

PHP Petri plate, stack size 10, 55 mm, Gamma irradiated, 10×112

 
Rs.20,550.00
   
1 * 1.120 items
 

High performance, gamma sterilized and disposable Petri dishes for every microbiology lab.

  • Manufactured in 10K cleanroom for dust free final products
  • Sterilized as per 10⁻⁶ gamma irradiation
  • Ensures highest possible flatness for zero wastage of media (A to B and C to D lesser than 0,01 mm)
  • Made of US FDA approved PS with best-in-class clarity and tensile strength
  • Rimming of plates are designed for better stacking, marking patch for writing

Manufactured under strict aseptic conditions, VWR® Petri plates are perfect for use in automated plate filing systems. The high transparency of these plates ensures accurate observation and measurement, reducing the margin for errors in research.

 : The innovative packaging of these PHP plates is breakage proof.

 : The innovative packaging of these PHP plates is breakage proof.

 
  • Ø:
    55 mm
  • Packed:
    10×112
  • Sterility:
    Gamma irradiated
  • Description:
    PHP Petri plate, stack size 10
  • Seq:
    08