BeneSphera™ E-21, ELISA microplate reader, multi-mode
BeneSphera™ E-21 ELISA Microplate Reader, Multi-mode
Catalog # BENP20001999
Supplier: BENESPHERA
Jump to:
Product Details & Documents
The BeneSphera™ E-21 ELISA reader is a bi-chromatic optic system with four wavelengths.
- 8-channel optic fibre system - get result in just 5 seconds
- Plate shaking feature
- Large touch screen LCD display
- Large memory to store 500 tests and 10000 results
- Ability to perform 12 different tests in one plate
- Multi-forms in results output including individual patient comprehensive report
Unit has optional built in modem for remote diagnosis and software upgrade.
Documents
Avantor BeneSphera™ Elisa plate Reader and Washer
Product Literature
Specifications
- Description:BeneSphera™ E-21 ELISA Microplate Reader, Multi-mode