Order Entry
India
ContactUsLinkComponent
Value Plastics® Sanitary to Hose Barb Adapter, 3/4" Tri-Clamp to ³/₈" ID Hose Barb, ADCF PP; 100/PK
Value Plastics® Sanitary to Hose Barb Adapter, 3/4" Tri-Clamp to ³/₈" ID Hose Barb, ADCF PP; 100/PK
Catalog # MFLA31515-89BLK
Value Plastics® Sanitary to Hose Barb Adapter, 3/4" Tri-Clamp to ³/₈" ID Hose Barb, ADCF PP; 100/PK
Catalog # MFLA31515-89BLK
About this item
Certificate of compliance included!
  • Animal derivative component free
  • Meet USP Class IV requirements
  • Sterilizable vis gamma irradiation
These sanitary fittings are the perfect to transition from sanitary clamp connections to seal or barb for flexible tubing. All adapters feature a radiused flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow—and no particulate build-up. There are no sharp edges in the flow path, so cleaning is very easy. Made from animal-derived ingredient-free polypropylene, these adapters offer good chemical resistance and may be sterilized by gamma irradiation. Material meets the requirements for USP Class VI.
Delivery: Includes certificate of compliance.
  • Barb size/tubing int.Ø (port 2):
    3/8"
  • Connection Type:
    Sanitary to Barb
  • Flow pattern:
    Straight
  • Sanitary clamp size (Port 1):
    3/4"
  • Description:
    Sanitary to Hose Barb Adapter, 3/4" Tri-Clamp to ³/₈" ID Hose Barb, ADCF PP; 100/PK
  • Seq:
    01
  • Environmentally Preferable:
  • Sterilisation:
    Ethylene oxide (ETO) or gamma
  • Pk:
    100
  • Fitting material:
    Polypropylene
  • Sanitary gasket size:
    3/4"
Frequently Bought Together