Order Entry
India
ContactUsLinkComponent
Safety Cap III GL45, Screw cap GL45 made of PP-black, insert made of PTFE with 3 capillary connections (UNF 1/4 ) and 1 connection f or air valve (UNF 1/4 ). Including 3 fittings for tubing O.D. 3,2 mm (PFA-black), 1 air valve with flag with note for 1 * 1 items
Catalog # 590-0028
Supplier:  VWR Collection
Safety Cap III GL45, Screw cap GL45 made of PP-black, insert made of PTFE with 3 capillary connections (UNF 1/4 ) and 1 connection f or air valve (UNF 1/4 ). Including 3 fittings for tubing O.D. 3,2 mm (PFA-black), 1 air valve with flag with note for 1 * 1 items
Catalog # 590-0028
Supplier:  VWR Collection
Frequently Bought Together