Order Entry
India
ContactUsLinkComponent
Value Plastics Color Coded Lock Ring (for Use With Ftllb or Ftlb Panel Mount Fittings), White Nylon
Value Plastics Color Coded Lock Ring (for Use With Ftllb or Ftlb Panel Mount Fittings), White Nylon
Catalog # MFLX45509-01
undefined
Value Plastics Color Coded Lock Ring (for Use With Ftllb or Ftlb Panel Mount Fittings), White Nylon
Catalog # MFLX45509-01
About this item
Premium luer fittings for critical applications.
  • Manufactured from 100% virgin raw materials in a Class 100,000 environment
  • Full material traceability on all items
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Manufactured in compliance with ISO 594-1 standards, these fittigns are inspected to be free from contaminants, and are fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity. Choose from a range of sizes, 1/16" to 1/4", with a variety of connection options. All fittings are sold in packs of 100.
  • Description:
    Color Coded Lock Ring (for Use With Ftllb or Ftlb Panel Mount Fittings), White Nylon
  • Seq:
    01
  • Environmentally Preferable:
  • Model:
    45509-01
  • Pk:
    1
  • Fitting material:
    Nylon
Frequently Bought Together