Value Plastics Barbed to sanitary adapter, 1/2, pack of 10
Catalog # MFLX31513-76
Supplier: Avantor Fluid Handling
About this item
All adapters feature a radiused flow surface controlled to 15 micron Ra to ensure a smooth, uninterrupted flow, no particulate build-up and easy cleaning.
Made from animal-derived ingredient-free polypropylene and PVDF, these adapters may be sterilized by gamma irradiation. Material meets the requirements for USP Class VI.
- Barb size/tubing int.Ø (port 2):½"
- Connection Type:Sanitary to Barb
- Flow pattern:Straight
- Sanitary clamp size (Port 1):1½"
- Description:Barbed to sanitary adapter, ½, pack of 10
- Environmentally Preferable:
- Sterilisation:Gamma only
- Pk:1
- Fitting material:Polypropylene
Frequently Bought Together