Masterflex® Male threaded adapter, 1/16" NPT, to 3/32" hose barb, polypropylene, pack of 10
Catalog # MFLX30641-01
Supplier: Avantor Fluid Handling
CE certificate
About this item
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.
- Flow pattern:Straight
- Description:Male threaded adapter, ¹⁄₁₆" NPT, to ³⁄₃₂" hose barb, polypropylene, pack of 10
- Seq:02
- Environmentally Preferable:
- Model:30641-01
- Sterilisation:Autoclave or ethylene oxide (ETO)
- Pk:10
- Fitting material:Polypropylene
Frequently Bought Together