Masterflex® Versatile Cap Adapter Insert, 83B, 2 x 1/4" ID Quick Connect Ports
Catalog # MFLX06058-52
Supplier: Avantor Fluid Handling
About this item
Interchangeable ports for flexibility.
- Choose from a variety of sizes and configurations
- Change the top connections of each carboy to match your application needs
- Autoclavable
Versatile carboy cap adapter inserts are made of autoclavable polypropylene or clear polycarbonate. Quick-connect hosebarb fittings are also available. Vent ports are 1/4" in diameter.
- Cap material:PP
- Cap Size:83B
- Description:Versatile Cap Adapter Insert, 83B, Autoclavable, 2 x 1/4" ID Quick Connect Ports
- Environmentally Preferable:
- Cap colour:White
- Pk:1
Frequently Bought Together