Masterflex® Fuse for FS-3400 and VS-4000 Centrifuges, 115 VAC
Catalog # MFLX83058-12
Supplier: Avantor Fluid Handling
About this item
Fixed or variable speed for 6 or 8 place rotors.
Centrifuges include a fixed angle rotor to spin tubes up to 10 ml. Choose from fixed or variable speed models; both models feature a 30-minute timer, suction cup rubber feet which firmly anchor them to the benchtop, and a stainless steel guard bowl to protect against spills. A safety lid latch cuts power to the motor if the lid is raised. The exterior is made of chemically resistant plastic.
What’s included: 6 place rotor, tube shield, 6-ft power cord.
- Description:Fuse for FS-3400 and VS-4000 Centrifuges, 115 VAC
- Seq:04
- Environmentally Preferable:
- Warranty:1 year
- Pk:1
Frequently Bought Together