Leeson 098000 NEMA Type 56C-face TEFC motor, 1/2 hp, 1750 rpm, 90 VDC
Catalog # MFLX70071-20
Supplier: Avantor Fluid Handling
CSA Certified
UL Listed
Specifications
See all product configurations- Catalog No:
- MFLX70071-20
- MFLX70074-10
- MFLX70071-40
- MFLX70073-00
- MFLX70074-00
- MFLX70071-50
- MFLX70072-20
- MFLX70071-10
- MFLX70071-30
- MFLX70072-10
- Motor:
- 0,45 kW
- 0,42 kW
- 0,9 kW
- 0,24 kW
- 0,32 kW
- 0,9 kW
- 0,32 kW
- 0,45 kW
- 0,45 kW
- 0,32 kW
- Motor enclosure type:
- TEFC
- XPRF
- TEFC
- WDN
- XPRF
- TEFC
- TENV
- TEFC
- TEFC
- TENV
- Power conduit box connection:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Shaft centerline:
- 8,26 cm (3,25")
- 8,26 cm (3,25")
- 8,26 cm (3,25")
- 8,26 cm (3,25")
- 8,26 cm (3,25")
- 8,26 cm (3,25")
- 8,26 cm (3,25")
- 8,26 cm (3,25")
- 8,26 cm (3,25")
- 8,26 cm (3,25")
- Shaft Ø:
- 1,59 cm (0,625")
- 1,59 cm (0,625")
- 1,59 cm (0,625")
- 1,59 cm (0,625")
- 1,59 cm (0,625")
- 1,59 cm (0,625")
- 1,59 cm (0,625")
- 1,59 cm (0,625")
- 1,59 cm (0,625")
- 1,59 cm (0,625")
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- Currently Viewing
Specifications
About this item
Low-cost DC motors available in numerous configurations to meet your needs.
- Power pulleys, belts, pumps, or other machinery
- Operate low-voltage models (12 or 24 VDC) with battery or solar power for fixed-speed applications
- Explosion-proof models are Class 1, Groups C & D; Class II, Groups F & G rated
DC motors are suited for a variety of environments.
Totally enclosed, fan-cooled (TEFC) motors utilize an external fan to circulate outside air to the motor for applications where the motor will be running continuously.
Totally enclosed, nonvented (TENV) motors are designed without an air opening to protect against dust and other debris.
Explosion-proof (XPRF) motors are approved for areas where hazardous materials are used.
Washdown (WDN) motors are suitable for applications requiring regular washing or sanitizing cycles.
To vary the speed on a DC motor, order a DC controller at the side.
- Power:90 VDC
- Description:NEMA Type 56C-face TEFC motor, 1/2 hp, 1750 rpm, 90 VDC
- Height:16,51 cm
- Length:30,48 cm
- Environmentally Preferable:
- Motor enclosure type:TEFC
- Power conduit box connection:Yes
- Shaft centerline:8,26 cm (3,25")
- Shaft Ø:1,59 cm (0,625")
- Motor:0,45 kW
- Warranty:1 year
- Pk:1
- Width:20,32 cm
- Max. motor RPM:1750 rpm
- Motor frame size:56C
- Motor hp:1/2
Frequently Bought Together