Labnet StripSpin Centrifuge Tube Adapter; 16x0.2 mL Tubes or Strips
Catalog # MFLX17707-11
Supplier: Avantor Fluid Handling
About this item
Choose the centrifuge tube adapter to suit your application needs!
- Expand and change the capacity of your centrifuge with these adapters
Customize your Labnet centrifuge with these tube adapters.
- Capacity:0,2 ml individual or strips
- Description:Centrifuge Tube Adapter; 16x0,2 mL Tubes or Strips
- Environmentally Preferable:
- Compatible with:Labnet "standard" size rotors
- Pk:1
Frequently Bought Together