Order Entry
India
ContactUsLinkComponent
Centurion Tube Supports for Short Centrifuge Tubes, 100 mL, 4/Bx
  MFLX17430-62
Centurion Tube Supports for Short Centrifuge Tubes, 100 mL, 4/Bx
  MFLX17430-62
Customize your centrifuge.
Get the most out of your centrifuge by adding buckets, tube adapters, tube supports, and a rolling base.
  • Description:
    Tube Supports for Short Centrifuge Tubes, 100 mL, 4/Bx
  • Environmentally Preferable:
  • Warranty:
    3 years
  • Compatible with:
    Centurion Heated Oil Centrifuge adapters 17430-42 and -43
  • Pk:
    1
Frequently Bought Together