Labnet C2400-SS Optional tube adapters, 16 x 0.2 mL, 1/pk
MFLX17410-50
Choose the centrifuge tube adapter to suit your application needs!
- Expand and change the capacity of your centrifuge with these adapters
Customize your Labnet centrifuge with these tube adapters.
- Description:Optional tube adapters, 16 x 0,2 mL, 1/pk
- Environmentally Preferable:
- Pk:1
- Warranty:1 year
Frequently Bought Together