Order Entry
India
ContactUsLinkComponent
Masterflex® Large Tube Adapters, 15 mL round bottom without cap, 4/pk
Masterflex® Large Tube Adapters, 15 mL round bottom without cap, 4/pk
Catalog # MFLX17406-58
Masterflex® Large Tube Adapters, 15 mL round bottom without cap, 4/pk
Catalog # MFLX17406-58

About this item

Expand and change the capacity of your centrifuge with these adapters.

  • Capacity:
    28 x 15 ml
  • Description:
    Large Tube Adapters, 15 mL round bottom without cap, 4/pk
  • Environmentally Preferable:
  • Model:
    17406-58
  • Compatible with:
    Cole-Parmer MP/MPR16, MP/MPR115 Centrifuges
  • Pk:
    4
  • Warranty:
    3 years
Frequently Bought Together