Order Entry
India
ContactUsLinkComponent
Masterflex® Medium Tube Adapters, 15 mL Falcon with conical tip and cap, 4/pk
Masterflex® Medium Tube Adapters, 15 mL Falcon with conical tip and cap, 4/pk
Catalog # MFLX17406-38
Masterflex® Medium Tube Adapters, 15 mL Falcon with conical tip and cap, 4/pk
Catalog # MFLX17406-38

About this item

Expand and change the capacity of your centrifuge with these adapters.

  • Capacity:
    8 x 15 ml
  • Description:
    Medium Tube Adapters, 15 mL Falcon with conical tip and cap, 4/pk
  • Environmentally Preferable:
  • Model:
    17406-38
  • Compatible with:
    Cole-Parmer MP0415, MP/MPR115 Centrifuges
  • Pk:
    4
  • Warranty:
    3 years
Frequently Bought Together