Masterflex® MP(R)115/MP(R)315 Centrifuge Microplate Rotor, 2 or 4 Plates
MFLX17406-06
Expand and change the capacity of your centrifuge with these rotors.
- Description:Centrifuge Microplate Rotor, 2 or 4 Plates
- Environmentally Preferable:
- Model:17406-06
- Pk:1
- Max. RPM:3500 rpm
- Min. RCF:2500 ×g
- Rotor type:Microplate
- Tube sizes accepted:96-well
- Warranty:3 years
Frequently Bought Together