Masterflex® Fitting, PVDF, Straight, Male Luer to Thread Adapter, 1/4-28 UNF; 25/PK
MFLX45513-82
CE certificate
Easily transition between luer fittings and UNF or NPT threads.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are male thread.
- Flow pattern:Straight
- Description:Adapter, PVDF, Male Luer To ¼-28 Thread, 25/pack 45513-82
- Seq:06
- Environmentally Preferable:
- Model:45513-82
- Connection type (port 1):Male luer lock
- Connection type (port 2):¼-28 UNF thread
- No. of ports:2
- Pk:25
- Fitting material:PVDF
Frequently Bought Together