Masterflex® Smart Clamp® Stainless Steel Sanitary Clamps, 2" Tri-Clamp; 1/EA
MFLX31608-91
Torque control for consistent sealing of fittings.
- Constructed of durable stainless steel
These sanitary clamps feature a unique design that allows fast clamping while reducing the repetitive strain injuries in hands and fingers that can occur with regular sanitary clamps; only a half turn is required. Clamps also have an adjustable torque for compression control and feature multiple settings for use with a variety of sanitary gaskets. Threadless design prevents soil and bacteria entrapment. Clamps also support ASME-BPE standards.
- Connection type:Sanitary
- Description:Smart Clamp® Stainless Steel Sanitary Clamps, 2" Tri-Clamp; 1/EA
- Material:Stainless steel
- Environmentally Preferable:
- Sanitary clamp size (port 1):2"
- Pk:1
- Clamp type:Sanitary
Frequently Bought Together