Parker Hannifin Fitting, PFA, Elbow, Compression to Thread Adapter, 3/4" OD x 3/4" NPT(M)
Catalog # MFLX31320-81
Supplier: Avantor Fluid Handling
CE certificate
About this item
Part of our vast assortment of compression fittings.
- Compression x NPT(M)
- Sterilized via autoclave or ethylene oxide (ETO)
- Constructed of durable PFA
Count on this this quality-made compression threaded adapter elbow to suit your needs.
- Connection Type:Compression to Thread
- Flow pattern:Elbow
- Thread size NPT(M) (port 2):¾"
- Description:Compression threaded adapter elbow, PFA, ¾" tubing OD x ¾" male NPT
- Environmentally Preferable:
- Sterilisation:Autoclave or ethylene oxide (ETO)
- Warranty:18 months
- Pk:1
- Fitting material:PFA
Frequently Bought Together