Micropump Inlet filter/strainer, 20-30 mesh, 1/2" barb
Catalog # MFLX75000-51
Supplier: Avantor Fluid Handling
About this item
Optimize performance of your Flux® centrifugal pump.
- Choose from a wide range of replacement parts and accessories
Replacement parts and accessories to keep you pumping.
- Description:Inlet filter/strainer, 20-30 mesh, 1/2" barb
- Environmentally Preferable:
- Warranty:1 year
- Compatible with:Multi-position double diaphragm pump
- Pk:1
Frequently Bought Together