Order Entry
India
ContactUsLinkComponent
Flowline LO10-1405 Optic Leak Detection Pp Sensor Long 12-36VDC
  MFLX43312-02
undefined
Flowline LO10-1405 Optic Leak Detection Pp Sensor Long 12-36VDC
  MFLX43312-02
  • Warranty:
  • 1 year
  • 1 year
  •  
Detects leaks in and around secondary containment areas.
  • Choose between polypropylene (PP) or PFA construction
The optic switch is an excellent choice for leak detection applications in and around the secondary containment vessel of sumps, tanks and pipes. The sensor can be applied in a broad range of liquids including sulfuric acid, jet fuel and de-ionized water. The relay output provides a reliable switch interface with remote devices such as a PLC, SCADA or alarm. For maximum flexibility, the sensor is offered in general purpose and intrinsically safe versions with two probe lengths and PP or PFA PTFE materials.
  • Description:
    Optic Leak Detection Pp Sensor Long 12-36VDC
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Pk:
    1
  • Power:
    12 - 36 VDC
Frequently Bought Together