Flowline Ultrasonic level switch; material, PP; mounting threads, 3/4" NPT (M)
MFLX43299-02
Compatible with Smart Trak and Switch-Pak installation fittings.
- CSA approved intrinsically safe for use in hazardous environments
- 60 VA relay is selectable as normally open or normally closed via power supply wiring polarity
The LU10 series ultrasonic level switch is an excellent choice for a broad range of liquids, including those with light coating or scaling-type characteristics. Liquid examples include xylene, chromic acid and light cooking oil.
- Max. pressure:150 psi
- Description:Ultrasonic level switch; material, PP; mounting threads, 3/4" NPT (M)
- Environmentally Preferable:
- Accuracy of depth range:±1 mm
- Max. process temperature:90 °C
- Min. process temperature:-40 °F
- Relay output:SPDT relay: 1 A at 60 VAC/VDC
- Warranty:1 year
- Pk:1
- Power:12 - 36 VDC
Frequently Bought Together