Order Entry
India
ContactUsLinkComponent
 
Hose Fittings, Band Clamps, for 2" OD Hose, 25/Pk
  MFLX33350-14
undefined
Hose Fittings, Band Clamps, for 2" OD Hose, 25/Pk
  MFLX33350-14

 

Quick and easy assembly without tools.

  • Constructed of durable stainless steel

Band Clamps are ideal for tightening hose over barbed fittings. Constructed of stainless steel. All band clamps require a band clamp tool to install.

  • Tubing ext.Ø:
    50,8 mm (2")
  • Description:
    Hose Fittings, Band Clamps, for 2" OD Hose, 25/Pk
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Pk:
    25
Frequently Bought Together