Parker Hannifin Flare reducing union, 1/2" x 3/8", PFA, Each
Catalog # MFLX30490-12
Supplier: Avantor Fluid Handling
About this item
Leak-tight and high-pressure connection for corrosive chemical applications.
- Constructed of durable PFA and PVDF
PFA wetted parts and PVDF nuts provide high purity and excellent chemical resistance. Leaf-free design withstands pressures to 225 psi. Ideal for high-vibration or side-load stressed applications. Low dead volume reduces particle entrapment, limits bacterial growth in deionized water, and simplifies rinsing.
- Connection Type:Flare Union
- Flow pattern:Straight
- Description:Flare reducing union, ½" x ⅜", PFA, Each
- Environmentally Preferable:
- Warranty:3 Months
- Pk:1
- Fitting material:PFA
Frequently Bought Together