Parker Hannifin Fitting, Stainless Steel, Tee, Compression Branch to Threaded Adapter, 1/4" OD x 1/4" OD x 1/4" NPT(M)
MFLX30486-64
Securely connect your rigid tubing.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
These branch tees feature a male NPT thread on the bottom branch for quick connection to a threaded port.
- Connection Type:Compression to Thread
- Flow pattern:T
- Description:Fitting, Stainless Steel, Tee, Compression Branch to Threaded Adapter, ¼" OD x ¼" OD x ¼" NPT(M)
- Environmentally Preferable:
- Sterilisation:Autoclave, ethylene oxide (ET0) or gamma
- Warranty:3 Months
- Pk:1
- Fitting material:Stainless Steel
Frequently Bought Together