Parker Hannifin Fittings, Stainless Steel, Straight, Compression Reducer, 3/8" OD x 1/4" OD
Catalog # MFLX30486-10
Supplier: Avantor Fluid Handling
About this item
Get a secure seal at higher pressures.
- Quick and easy assembly without tools
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
- Connection Type:Compression Reducer
- Flow pattern:Straight
- Description:Reducing union, ⅜" × ¼", Stainless Steel, Each
- Environmentally Preferable:
- Warranty:3 Months
- Pk:1
- Fitting material:Stainless Steel
Frequently Bought Together