Precision Digital PD6820-0K1 Explosion-Proof Rate/Totalizer, Input Loop Powered
MFLX30004-66
CE Compliant
Program your process meter on-site.
- 5-digit rate and 7-character total/tag display
- Backlight standard
- Rotatable display
- FM, CSA, ATEX, IECEx, CE approved
The ProtEX-RTs are rugged, explosion-proof, full-featured rate/totalizers designed for demanding applications in hazardous areas or in the harshest environmental conditions. The PD6820 accepts analog inputs and the PD6830 accepts pulse inputs. They are programmed using the four SafeTouch™ through-glass buttons without removing the cover, or the four mechanical buttons. The sleek die-cast aluminum enclosure is provided with two ¾-inch threaded conduit holes and integrated pipe or wall mounting flanges.
- Display type:0,7" LED; 0,4" LED
- Description:Explosion-Proof Rate/Totalizer, Input Loop Powered
- Environmentally Preferable:
- Control input:4 - 20 mA
- Display digits:5; 7
- Enclosure:NEMA 4X, 7, 9; IP68; explosion-proof die-cast aluminum
- Min. operating temperature:–40 °F
- Overall depth:123,8 mm (4,875")
- Overall height:133,4 mm (5,25")
- Max. operating temperature:167 °F
- Overall width:142,9 mm (5,625")
- Warranty:3 years
- Pk:1
Frequently Bought Together