Filter Cartridge, Pleated Polyester, 20 micron nominal-rated, 19-1/2" L
MFLX29808-29
Pleated filter gives you more surface area for higher flowrate and longer life.
- Constructed of pleated polyester
Pleated Polyester Cartridges are general purpose cartridges offering long life and high dirt holding capacity. Polyester cartridges are cleanable and reusable in most applications and are NSF 61 listed.
- Filter media:Polyester
- Micron Rating:20
- Description:Filter Cartridge, Pleated Polyester, 20 micron nominal-rated, 19-1/2" L
- Material:Polyester
- Environmentally Preferable:
- Warranty:1 year
- Pk:1
- Cartridge length:19,5"
- Filter length:19,5"
Frequently Bought Together