Replacement Cores for Sanitary Strainers
Side-Inlet Sanitary Strainer Core; 1/4" Perforations
Catalog # MFLX29590-72
Supplier: Avantor Fluid Handling
Jump to:
Product Details & Documents
Protect products and expensive process equipment from coarse or fine particulates.
- 316 SS fluid contact surfaces are polished to a smooth, blemish free finish
- Standard clamp sizes allow easy installation or retrofit
- Side-inlet design lets you clean housing and change screens without disconnecting the strainer from your process line
Sanitary strainers are ideal for particulate removal or pre-filtration in the processing of foods, beverages, pharmaceuticals, cosmetics, and other critical compounds. Housings are available in in-line and side-inlet configurations.
Delivery information: Strainer assemblies include housing, perforated core, internal cap and spring, assembly clamp and gasket. Order wire mesh overscreens of the desired mesh size separately.
Specifications
- Description:Side-Inlet Sanitary Strainer Core; 1/4" Perforations
- Cat. No.:MFLX29590-72
- Filter type:Strainer