Order Entry
India
ContactUsLinkComponent
Hermle 12 x 15 mL Angle Rotor (32°)
Hermle 12 x 15 mL Angle Rotor (32°)
Catalog # MFLX51705-58
Hermle 12 x 15 mL Angle Rotor (32°)
Catalog # MFLX51705-58

About this item

Versatile rotors for your compact centrifuge.

  • Choose from six 50 ml tubes or twelve 15 ml tubes
  • Description:
    12 x 15 mL Angle Rotor (32°)
  • Environmentally Preferable:
  • Pk:
    1
  • Warranty:
    3 years
Frequently Bought Together