Masterflex® Short tube adapters, for 4 to 7 mL tubes, 10/pk
Catalog # MFLX83058-08
Supplier: Avantor Fluid Handling
About this item
Use tube adapters in centrifuge rotors and buckets to accommodate your tube size.
- Capacity:4 - 7 ml
- Description:Short tube adapters, for 4 to 7 mL tubes, 10/pk
- Environmentally Preferable:
- Model:83058-08
- Pk:10
- Warranty:1 year
Frequently Bought Together