- Catalog No:
- MFLX17701-30
- MFLX17701-40
- MFLX17701-29
- Tube capacity:
- 4 x 145 mL
- Bucket is sold separately-4 x standard microplates or -2 x deepwell microplates
- 18 x 5 mL
- Max. speed:
- 4500 rpm
- 4400 rpm
- 5500 rpm
- Warranty:
- 3 years
- 3 years
- 1 year
Exceptional capacity and flexible processing adds value to any lab.
- SMARTSpin imbalance detection system prevents hazardous sample running
- Quality design and brushless drive ensure years of quiet use
- Biocontainment option keeps you safe
Conforming to the latest safety standards, the Megafuge 8 is a compact bench centrifuge that satisfies many sample running options. The secure push-button Auto-Lock® rotor exchange system with glove-friendly design allows you to change out rotors with ease, requiring only one hand! The SMARTSpin imbalance detection system optimizes acceleration, braking, and load imbalance for smooth runs and reproducible separations.
Centrifuge features one-touch operation and pre-saved programming with password protection. The control panel with highly visible backlit display is glove- and detergent-friendly. Brushless motor provides quiet performance and a stress free work environment.
Rotors require adapters, order separately.
- Max. speed:4500 rpm
- Description:SwingingBucketRotor,4x145mL,unsealed,requiresadapter
- Environmentally Preferable:
- Pk:1
- Tube capacity:4 x 145 mL
- Warranty:3 years