Masterflex® Centrifuge Tube Adapters, Small for 1.5to3 mL Tubes; 2/Pk
MFLX17415-26
Use tube adapters in centrifuge rotors and buckets to accommodate your tube size.
- Capacity:1,5 - 3 ml
- Description:Centrifuge Tube Adapters,Small, for 1,5 to 3 mLTubes; 2/Pk
- Environmentally Preferable:
- Model:17415-26
- Compatible with:17415 Series
- Pk:2
- Warranty:1 year
Frequently Bought Together