Masterflex® Centrifuge Power Adapter, 12 VDC - 110 VAC
Catalog # MFLX17415-22
Supplier: Avantor Fluid Handling
About this item
Convenient power adapters and replacement fuses to keep you spinning.
- Description:Centrifuge Power Adapter, 12 VDC to 110 VAC
- Environmentally Preferable:
- Warranty:1 year
- Compatible with:17415 Series - Power cord (110 VAC)
- Pk:1
- Power:110 VAC
Frequently Bought Together