Order Entry
India
ContactUsLinkComponent
Masterflex® Optional Tube Adapter for 5 to 15 mL tubes (for 17414-series centrifuges)
Masterflex® Optional Tube Adapter for 5 to 15 mL tubes (for 17414-series centrifuges)
Catalog # MFLX17414-31
Masterflex® Optional Tube Adapter for 5 to 15 mL tubes (for 17414-series centrifuges)
Catalog # MFLX17414-31

About this item

Use tube adapters in centrifuge rotors and buckets to accommodate your tube size.

  • Capacity:
    5 - 15 ml
  • Description:
    Optional Tube Adapter for 5 to 15 mL tubes (for 17414-series centrifuges)
  • Environmentally Preferable:
  • Model:
    17414-31
  • Compatible with:
    17414 Series
  • Pk:
    2
  • Warranty:
    1 year
Frequently Bought Together