Order Entry
India
ContactUsLinkComponent
 
Masterflex® Large Tube Adapters, 15 mL Falcon w/ conical tip and cap, 4/pk
  MFLX17406-61
Masterflex® Large Tube Adapters, 15 mL Falcon w/ conical tip and cap, 4/pk
  MFLX17406-61

 

Expand and change the capacity of your centrifuge with these adapters.

  • Capacity:
    16 x 15 ml
  • Description:
    Large Tube Adapters, 15 mL Falcon w/ conical tip and cap, 4/pk
  • Environmentally Preferable:
  • Model:
    17406-61
  • Compatible with:
    Cole-Parmer MP/MPR16, MP/MPR115 Centrifuges
  • Pk:
    4
  • Warranty:
    3 years
Frequently Bought Together