Order Entry
India
ContactUsLinkComponent
 
Eppendorf 22 63 740-1 Swing-bucket rotor; four x 100 mL buckets
  MFLX02750-74
Eppendorf 22 63 740-1 Swing-bucket rotor; four x 100 mL buckets
  MFLX02750-74

 

Expand or change your centrifuge capacity with the rotors.

  • Description:
    Swing-bucket rotor; four x 100 mL buckets
  • Environmentally Preferable:
  • Pk:
    1
  • No. of tubes accepted:
    4
  • Rotor type:
    Swing-out
  • Tube sizes accepted:
    100 ml
  • Max. RCF:
    4500 ×g
  • Warranty:
    2 years
Frequently Bought Together